3-Bromo-4-methylpyridine, 97%, Thermo Scientific Chemicals
3-Bromo-4-methylpyridine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-4-methylpyridine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 392320010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3430-22-6
IUPAC Name3-bromo-4-methylpyridine
Molecular FormulaC6H6BrN
InChI KeyGSQZOLXWFQQJHJ-UHFFFAOYSA-N
SMILESCC1=CC=NC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5600 to 1.5640 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only