Thermo Scientific Chemicals

Propionyl bromide, 95%, Thermo Scientific Chemicals

Catalog number: 392360250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Propionyl bromide, 95%, Thermo Scientific Chemicals

Catalog number: 392360250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392360250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
598-22-1
IUPAC Name
propanoyl bromide
Molecular Formula
C3H5BrO
InChI Key
RIBFXMJCUYXJDZ-UHFFFAOYSA-N
SMILES
CCC(Br)=O
Infrared spectrum
Conforms
Refractive index
1.4525 to 1.4575 (20°C, 589 nm)
GC
>=94 %
Appearance (Color)
Colorless to yellow
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text