4-Iodobiphenyl, 97+%, Thermo Scientific Chemicals
4-Iodobiphenyl, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Iodobiphenyl, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 392390010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-iodo-1,1'-biphenyl
Molecular FormulaC12H9I
InChI KeyNXYICUMSYKIABQ-UHFFFAOYSA-N
SMILESIC1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol)280.11
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point110°C to 114°C
Infrared spectrumConforms
Appearance (Color)Off-white
HPLC>=97.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only