3-Hydroxyphenylacetylene, 95+%, Thermo Scientific Chemicals
3-Hydroxyphenylacetylene, 95+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Hydroxyphenylacetylene, 95+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 392410050
Price (USD)/ Each
343.00
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
343.00
Add to cart
3-Hydroxyphenylacetylene, 95+%, Thermo Scientific Chemicals
Catalog number392410050
Price (USD)/ Each
343.00
-
Add to cart
Chemical Identifiers
IUPAC Name3-ethynylphenol
Molecular FormulaC8H6O
InChI KeyAODMJIOEGCBUQL-UHFFFAOYSA-N
SMILESOC1=CC=CC(=C1)C#C
Molecular Weight (g/mol)118.14
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=95.0 %
Refractive index1.5840 to 1.5880 (20°C, 589 nm)
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only