Thermo Scientific Chemicals

Methoxytrimethylsilane, 96+%, Thermo Scientific Chemicals

Catalog number: 392470250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Methoxytrimethylsilane, 96+%, Thermo Scientific Chemicals

Catalog number: 392470250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392470250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1825-61-2
IUPAC Name
methoxytrimethylsilane
Molecular Formula
C4H12OSi
InChI Key
POPACFLNWGUDSR-UHFFFAOYSA-N
SMILES
CO[Si](C)(C)C
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Refractive index
1.3660 to 1.3680 (20°C, 589 nm)
GC
>=96.0 %
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text