p-Tolylacetylene, 97%, Thermo Scientific Chemicals
p-Tolylacetylene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

p-Tolylacetylene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 392560050
Price (USD)/ Each
72.30
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
72.30
Add to cart
p-Tolylacetylene, 97%, Thermo Scientific Chemicals
Catalog number392560050
Price (USD)/ Each
72.30
-
Add to cart
Chemical Identifiers
IUPAC Name1-ethynyl-4-methylbenzene
Molecular FormulaC9H8
InChI KeyKSZVOXHGCKKOLL-UHFFFAOYSA-N
SMILESCC1=CC=C(C=C1)C#C
Molecular Weight (g/mol)116.16
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.5460 to 1.5480 (20°C, 589 nm)
Appearance (Color)Light yellow to light brown
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only