Thermo Scientific Chemicals

1-Bromoisoquinoline, 95+%, Thermo Scientific Chemicals

Catalog number: 392590010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Bromoisoquinoline, 95+%, Thermo Scientific Chemicals

Catalog number: 392590010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392590010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1532-71-4
IUPAC Name
1-bromoisoquinoline
Molecular Formula
C9H6BrN
InChI Key
YWWZASFPWWPUBN-UHFFFAOYSA-N
SMILES
BrC1=C2C=CC=CC2=CC=N1
Melting point
42°C to 48°C
Appearance (Color)
Yellow to brown
Appearance (Form)
Powder and/or chunks
Infrared spectrum
Conforms
HPLC
>=95.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text