Thermo Scientific Chemicals

2-Cyclohexylethyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: 392640250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Cyclohexylethyl bromide, 98%, Thermo Scientific Chemicals

Catalog number: 392640250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392640250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1647-26-3
IUPAC Name
(2-bromoethyl)cyclohexane
Molecular Formula
C8H15Br
InChI Key
JRQAAYVLPPGEHT-UHFFFAOYSA-N
SMILES
BrCCC1CCCCC1
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.4890 to 1.4910 (20°C, 589 nm)
GC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text