Methyl 6-bromohexanoate, 96+%, Thermo Scientific Chemicals
Methyl 6-bromohexanoate, 96+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 6-bromohexanoate, 96+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number 392690250
Price (USD)/ Each
130.00
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
130.00
Add to cart
Methyl 6-bromohexanoate, 96+%, Thermo Scientific Chemicals
Catalog number392690250
Price (USD)/ Each
130.00
-
Add to cart
Chemical Identifiers
CAS14273-90-6
IUPAC Namemethyl 6-bromohexanoate
Molecular FormulaC7H13BrO2
InChI KeyKYLVAMSNNZMHSX-UHFFFAOYSA-N
SMILESCOC(=O)CCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4630 to 1.4650 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only