Thermo Scientific Chemicals

4-(Chloromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 392890500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Chloromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 392890500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392890500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1642-81-5
IUPAC Name
4-(chloromethyl)benzoic acid
Molecular Formula
C8H7ClO2
InChI Key
OITNBJHJJGMFBN-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC=C(CCl)C=C1
Titration with NaOH
>=97.5 %
Infrared spectrum
Conforms
HPLC
>=97.5 %
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text