Thermo Scientific Chemicals

4-(Bromomethyl)phenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 392900050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

4-(Bromomethyl)phenylacetic acid, 97%, Thermo Scientific Chemicals

Catalog number: 392900050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392900050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13737-36-5
IUPAC Name
2-[4-(bromomethyl)phenyl]acetic acid
Molecular Formula
C9H9BrO2
InChI Key
WCOCCXZFEJGHTC-UHFFFAOYSA-N
SMILES
OC(=O)CC1=CC=C(CBr)C=C1
Titration with NaOH
>=96.0 %
HPLC
>=96.0 %
Infrared spectrum
Conforms
Appearance (Color)
White to light beige
Appearance (Form)
Crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text