Thermo Scientific Chemicals

Methanesulfinic acid, sodium salt, 85%, Thermo Scientific Chemicals

Catalog number: 392960100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Methanesulfinic acid, sodium salt, 85%, Thermo Scientific Chemicals

Catalog number: 392960100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 392960100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
20277-69-4
IUPAC Name
sodium methanesulfinate
Molecular Formula
CH3NaO2S
InChI Key
LYPGDCWPTHTUDO-UHFFFAOYSA-M
SMILES
[Na+].CS([O-])=O
Appearance (Color)
White to light beige
Appearance (Form)
Powder
Titration with HClO4
>=80 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text