Thermo Scientific Chemicals

3-Methoxybenzyl bromide, 98+%, Thermo Scientific Chemicals

Catalog number: 393110250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Methoxybenzyl bromide, 98+%, Thermo Scientific Chemicals

Catalog number: 393110250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 393110250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
874-98-6
IUPAC Name
1-(bromomethyl)-3-methoxybenzene
Molecular Formula
C8H9BrO
InChI Key
ZKSOJQDNSNJIQW-UHFFFAOYSA-N
SMILES
COC1=CC=CC(CBr)=C1
GC
>=98.0 %
Appearance (Color)
Clear colorless to light yellow
Refractive index
1.5740 to 1.5760 (20°C, 589 nm)
Infrared spectrum
Conforms
Appearance (Form)
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text