Ammonium chloride, for analysis, Thermo Scientific Chemicals
Ammonium chloride, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium chloride, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 393182500
Price (USD)/ Each
37.65
Online exclusive
41.80 
Save 4.15 (10%)
-
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
41.80 
Save 4.15 (10%)
Add to cart
Ammonium chloride, for analysis, Thermo Scientific Chemicals
Catalog number393182500
Price (USD)/ Each
37.65
Online exclusive
41.80 
Save 4.15 (10%)
-
Add to cart
Chemical Identifiers
CAS12125-02-9
IUPAC Nameamine hydrochloride
Molecular FormulaClH4N
InChI KeyNLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILESN.Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration Argentometric>=99.0 %
Insoluble matter=<0.01 % (5 % in water)
Residue after ign. (sulfated)=<0.01 %
Heavy metals (as Pb)=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only