Ammonium fluoride, 98+%, for analysis, Thermo Scientific Chemicals
Ammonium fluoride, 98+%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium fluoride, 98+%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Plastic bottle
Catalog number 393192500
Price (USD)/ Each
75.65
Online exclusive
83.50 
Save 7.85 (9%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
75.65
Online exclusive
83.50 
Save 7.85 (9%)
Add to cart
Ammonium fluoride, 98+%, for analysis, Thermo Scientific Chemicals
Catalog number393192500
Price (USD)/ Each
75.65
Online exclusive
83.50 
Save 7.85 (9%)
-
Add to cart
Chemical Identifiers
CAS12125-01-8
IUPAC Nameammonium hydrogen difluoride
Molecular FormulaF2H5N
InChI KeyKVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILES[H+].[NH4+].[F-].[F-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess NaOH>=98.0 %
Sulfated ash=<0.01 %
(NH4)HF2=<1.0 %
Appearance (Form)Adhering crystalline powder
Insoluble matter=<0.005 % (in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only