Ammonium bicarbonate, 99%, for analysis, Thermo Scientific Chemicals
Ammonium bicarbonate, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium bicarbonate, 99%, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Plastic bottle
Catalog number 393210010
Price (USD)/ Each
82.65
Online exclusive
91.80 
Save 9.15 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
82.65
Online exclusive
91.80 
Save 9.15 (10%)
Add to cart
Ammonium bicarbonate, 99%, for analysis, Thermo Scientific Chemicals
Catalog number393210010
Price (USD)/ Each
82.65
Online exclusive
91.80 
Save 9.15 (10%)
-
Add to cart
Chemical Identifiers
CAS1066-33-7
IUPAC Namedizinc(2+) diammonium tricarbonate
Molecular FormulaC3H8N2O9Zn2
InChI KeyQOPSSYZLVIENBR-UHFFFAOYSA-J
SMILES[NH4+].[NH4+].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<5 ppm
Potassium (K)=<10 ppm
Sodium (Na)=<20 ppm
Heavy metals (as Pb)=<5 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only