2-(4-Bromophenyl)-1,3-dioxolane, 98+%, Thermo Scientific Chemicals
2-(4-Bromophenyl)-1,3-dioxolane, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(4-Bromophenyl)-1,3-dioxolane, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 393220010
Price (USD)/ Each
72.60
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
72.60
Add to cart
2-(4-Bromophenyl)-1,3-dioxolane, 98+%, Thermo Scientific Chemicals
Catalog number393220010
Price (USD)/ Each
72.60
-
Add to cart
Chemical Identifiers
CAS10602-01-4
IUPAC Name2-(4-bromophenyl)-1,3-dioxolane
Molecular FormulaC9H9BrO2
InChI KeyZYIMHOWVWWHLDN-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C1OCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
Refractive index1.5630 to 1.5650 (20°C, 589 nm)
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only