Sand, for analysis, 40-100 mesh
Sand, for analysis, 40-100 mesh
Sand, for analysis, 40-100 mesh
Sand, for analysis, 40-100 mesh
Thermo Scientific Chemicals

Sand, for analysis, 40-100 mesh

CAS: 14808-60-7 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 393300050
Price (USD)
408.65
Special offer
Online exclusive
480.00
Save 71.35 (15%)
Each
-
Add to cart
Quantity:
5 kg
Packaging:
Plastic bottle
Recurring order eligible. Learn more »
Request bulk or custom format
Price (USD)
408.65
Special offer
Online exclusive
480.00
Save 71.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystals
Particle size(D50) 205 µm (Typical)
Silicon dioxide (SiO2)>=99.80 % (Typical)
Calcium (Ca)=<80 ppm (Typical)
Chromium (Cr)=<0.5 ppm (Typical)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0469460Certificate of AnalysisJan 25, 2025393300050, 393302500, 393300010
A0460591Certificate of AnalysisFeb 22, 2024393300050, 393302500, 393300010
A0451222Certificate of AnalysisMay 02, 2023393300050, 393302500, 393300010
A0393276Certificate of AnalysisDec 14, 2022393300050, 393302500, 393300010
A0412184Certificate of AnalysisDec 13, 2022393300050, 393302500, 393300010
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection