Sand, for analysis, 40-100 mesh, Thermo Scientific Chemicals
Sand, for analysis, 40-100 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sand, for analysis, 40-100 mesh, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 393302500
Price (USD)/ Each
64.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
76.20 
Save 11.55 (15%)
-
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
64.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
76.20 
Save 11.55 (15%)
Add to cart
Sand, for analysis, 40-100 mesh, Thermo Scientific Chemicals
Catalog number393302500
Price (USD)/ Each
64.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
76.20 
Save 11.55 (15%)
-
Add to cart
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige to grey
Iron (Fe)=<65 ppm (Typical)
Aluminium (Al)=<250 ppm (Typical)
Titanium (Ti)=<150 ppm (Typical)
Potassium (K)=<50 ppm (Typical)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only