Thermo Scientific Chemicals

DL-Alanine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 393311000
100 g, Glass Bottle, Each
Thermo Scientific Chemicals

DL-Alanine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: 393311000
100 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 393311000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13515-97-4
IUPAC Name
methyl 2-aminopropanoate hydrochloride
Molecular Formula
C4H10ClNO2
InChI Key
IYUKFAFDFHZKPI-UHFFFAOYNA-N
SMILES
Cl.COC(=O)C(C)N
Titration with HClO4
>=98.0 %
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text