Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydroxide, 1N solution in ethanol, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Plastic bottle
Catalog number 393322500
Price (USD)/ Each
-
Quantity:
250 mL
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS1310-58-3, 64-17-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless to yellow to brown
Strength0.950 to 1.050 N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use