Thermo Scientific Chemicals

(S)-4-(1-Hydroxyethyl)pyridine, 99+%, (99+% ee), Thermo Scientific Chemicals

Catalog number: 393370010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(S)-4-(1-Hydroxyethyl)pyridine, 99+%, (99+% ee), Thermo Scientific Chemicals

Catalog number: 393370010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 393370010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
54656-96-1
IUPAC Name
1-(pyridin-4-yl)ethan-1-ol
Molecular Formula
C7H9NO
InChI Key
HVOAMIOKNARIMR-UHFFFAOYNA-N
SMILES
CC(O)C1=CC=NC=C1
Appearance (Form)
Powder or crystals and/or chunks
Appearance (Color)
White to cream
Infrared spectrum
Conforms
Melting point
67°C to 70°C
Specific optical rotation
-56° to -61° (20°C, 589 nm) (c=1, chloroform)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text