1-Bromobenzyl methyl ether, 98%, Thermo Scientific Chemicals
1-Bromobenzyl methyl ether, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Bromobenzyl methyl ether, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 393400050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52711-30-5
IUPAC Name1-bromo-2-(methoxymethyl)benzene
Molecular FormulaC8H9BrO
InChI KeyQFAZLCRHLRJNAW-UHFFFAOYSA-N
SMILESCOCC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Refractive index1.5470 to 1.5500 (20°C, 589 nm)
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only