Thermo Scientific Chemicals

1-Bromobenzyl methyl ether, 98%, Thermo Scientific Chemicals

Catalog number: 393400250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

1-Bromobenzyl methyl ether, 98%, Thermo Scientific Chemicals

Catalog number: 393400250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 393400250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52711-30-5
IUPAC Name
1-bromo-2-(methoxymethyl)benzene
Molecular Formula
C8H9BrO
InChI Key
QFAZLCRHLRJNAW-UHFFFAOYSA-N
SMILES
COCC1=CC=CC=C1Br
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Liquid
Refractive index
1.5470 to 1.5500 (20°C, 589 nm)
GC
>=97.5 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text