Thermo Scientific Chemicals

Butyraldehyde diethylacetal, 97%, Thermo Scientific Chemicals

Catalog number: 393431000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Butyraldehyde diethylacetal, 97%, Thermo Scientific Chemicals

Catalog number: 393431000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 393431000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3658-95-5
IUPAC Name
1,1-diethoxybutane
Molecular Formula
C8H18O2
InChI Key
UVHXZFGCCJLFMX-UHFFFAOYSA-N
SMILES
CCCC(OCC)OCC
Appearance (Form)
Liquid
Infrared spectrum
Conforms
GC
>=96.0 %
Refractive index
1.3950 to 1.3980 (20°C, 589 nm)
Appearance (Color)
Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text