Caffeine, 1mg/ml in methanol, Thermo Scientific Chemicals
Caffeine, 1mg/ml in methanol, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Caffeine, 1mg/ml in methanol, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 mL
Packaging:
Glass bottle
Catalog number 393540010
Price (USD)/ Each
56.65
Online exclusive
66.60 
Save 9.95 (15%)
-
Add to cart
Quantity:
1 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
66.60 
Save 9.95 (15%)
Add to cart
Caffeine, 1mg/ml in methanol, Thermo Scientific Chemicals
Catalog number393540010
Price (USD)/ Each
56.65
Online exclusive
66.60 
Save 9.95 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name1,3,7-trimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC8H10N4O2
InChI KeyRYYVLZVUVIJVGH-UHFFFAOYSA-N
SMILESCN1C=NC2=C1C(=O)N(C)C(=O)N2C
Molecular Weight (g/mol)194.19
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear colorless
Strength0.95 to 1.05 mg/ml
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only