Quartz Wool
Thermo Scientific Chemicals

Quartz Wool

Filter aid
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Catalog number 393611000
Price (USD)/ Each
234.65
Online exclusive
260.00 
Save 25.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
234.65
Online exclusive
260.00 
Save 25.35 (10%)
Add to cart
Quartz Wool
Catalog number393611000
Price (USD)/ Each
234.65
Online exclusive
260.00 
Save 25.35 (10%)
-
Add to cart
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Coarse fibers
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only