2-Bromo-alpha-methylbenzyl alcohol, 99%, Thermo Scientific Chemicals
2-Bromo-alpha-methylbenzyl alcohol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-alpha-methylbenzyl alcohol, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 393640010
Price (USD)/ Each
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5411-56-3
IUPAC Name1-(2-bromophenyl)ethan-1-ol
Molecular FormulaC8H9BrO
InChI KeyDZLZSFZSPIUINR-UHFFFAOYNA-N
SMILESCC(O)C1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.5590 to 1.5610 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only