Thermo Scientific Chemicals

2-Bromo-alpha-methylbenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 393640250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

2-Bromo-alpha-methylbenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog number: 393640250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 393640250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5411-56-3
IUPAC Name
1-(2-bromophenyl)ethan-1-ol
Molecular Formula
C8H9BrO
InChI Key
DZLZSFZSPIUINR-UHFFFAOYNA-N
SMILES
CC(O)C1=CC=CC=C1Br
Appearance (Form)
Liquid
Refractive index
1.5590 to 1.5610 (20°C, 589 nm)
Appearance (Color)
Clear yellow
Infrared spectrum
Conforms
GC
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text