1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea, 95%, Thermo Scientific Chemicals
1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3-Bis(tert-butoxycarbonyl)-2-methyl-2-thiopseudourea, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass Bottle
Catalog number 393740250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS107819-90-9
IUPAC Nametert-butyl N-({[(tert-butoxy)carbonyl]amino}(methylsulfanyl)methylidene)carbamate
Molecular FormulaC12H22N2O4S
InChI KeyUQJXXWHAJKRDKY-UHFFFAOYSA-N
SMILESCSC(NC(=O)OC(C)(C)C)=NC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
HPLC>=94 %
Infrared spectrumConforms
Appearance (Color)White to beige
Melting point118°C to 125°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only