1-Butanol, for HPLC
1-Butanol, for HPLC
1-Butanol, for HPLC
Thermo Scientific Chemicals

1-Butanol, for HPLC

CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 393750010
Price (USD)
101.65
Special offer
Online exclusive
Ends: 26-Sep-2025
119.00
Save 17.35 (15%)
Each
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
101.65
Special offer
Online exclusive
Ends: 26-Sep-2025
119.00
Save 17.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.0003 %
Infrared spectrumConforms
Water=<0.05 %
UV1 cm cell vs HPLC-grade water
UVat 220 nm A: =<0.301
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only

Share catalog number, name or link