Thermo Scientific Chemicals

4-Methoxy-2-methylphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 393850010
1 g, Glass Bottle, Each
Thermo Scientific Chemicals

4-Methoxy-2-methylphenylboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: 393850010
1 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 393850010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
208399-66-0
IUPAC Name
(4-methoxy-2-methylphenyl)boronic acid
Molecular Formula
C8H11BO3
InChI Key
AMSQNQJCBXQYEX-UHFFFAOYSA-N
SMILES
COC1=CC=C(B(O)O)C(C)=C1
Appearance (Color)
White to off-white
Infrared spectrum
Conforms
HPLC
>=96.0 %
Appearance (Form)
Crystalline solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text