Sodium tripolyphosphate, for analysis, Thermo Scientific Chemicals
Sodium tripolyphosphate, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium tripolyphosphate, for analysis, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 393960250
Price (USD)/ Each
29.65
Online exclusive
32.60 
Save 2.95 (9%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
29.65
Online exclusive
32.60 
Save 2.95 (9%)
Add to cart
Sodium tripolyphosphate, for analysis, Thermo Scientific Chemicals
Catalog number393960250
Price (USD)/ Each
29.65
Online exclusive
32.60 
Save 2.95 (9%)
-
Add to cart
Chemical Identifiers
CAS7758-29-4
IUPAC Namepentasodium bis(phosphonatooxy)phosphinate
Molecular FormulaNa5O10P3
InChI KeyHWGNBUXHKFFFIH-UHFFFAOYSA-I
SMILES[Na+].[Na+].[Na+].[Na+].[Na+].[O-]P([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration>=98.0 %
Nickel (Ni)=<50 ppm
Appearance (Form)Powder
Cadmium (Cd)=<50 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only