Thermo Scientific Chemicals

3-Benzyloxyphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 394020010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

3-Benzyloxyphenylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: 394020010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 394020010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
59146-68-8
IUPAC Name
[3-(benzyloxy)phenyl]hydrazine hydrochloride
Molecular Formula
C13H15ClN2O
InChI Key
MMMGCMOISVZVKZ-UHFFFAOYSA-N
SMILES
Cl.NNC1=CC(OCC2=CC=CC=C2)=CC=C1
Appearance (Color)
Violet
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text