2-Iodophenylacetic acid, 97+%, Thermo Scientific Chemicals
2-Iodophenylacetic acid, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Iodophenylacetic acid, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 394040050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18698-96-9
IUPAC Name2-(2-iodophenyl)acetic acid
Molecular FormulaC8H7IO2
InChI KeyIUHXGZHKSYYDIL-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.0 %
Appearance (Color)White to cream to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Melting point113°C to 118°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only