Tetrabutylammonium hydrogen sulfate, 99%, extra pure, Thermo Scientific Chemicals
Tetrabutylammonium hydrogen sulfate, 99%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrabutylammonium hydrogen sulfate, 99%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 394200050
Price (USD)/ Each
24.70
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
24.70
Add to cart
Tetrabutylammonium hydrogen sulfate, 99%, extra pure, Thermo Scientific Chemicals
Catalog number394200050
Price (USD)/ Each
24.70
-
Add to cart
Chemical Identifiers
CAS32503-27-8
IUPAC Nametetrabutylazanium hydrogen sulfate
Molecular FormulaC16H37NO4S
InChI KeySHFJWMWCIHQNCP-UHFFFAOYSA-M
SMILESOS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point169°C to 173°C
Titration with NaOH>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only