1x1 image pixel for data collection
Tetrabutylammonium hydrogen sulfate, 99%, extra pure
Tetrabutylammonium hydrogen sulfate, 99%, extra pure
Tetrabutylammonium hydrogen sulfate, 99%, extra pure
Tetrabutylammonium hydrogen sulfate, 99%, extra pure
Thermo Scientific Chemicals

Tetrabutylammonium hydrogen sulfate, 99%, extra pure

CAS: 32503-27-8 | C16H37NO4S | 339.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 394205000
Price (USD)
309.65
Special offer
Online exclusive
Ends: 26-Sep-2025
382.00
Save 72.35 (19%)
Each
In stock
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
309.65
Special offer
Online exclusive
Ends: 26-Sep-2025
382.00
Save 72.35 (19%)
Each
Add to cart
Chemical Identifiers
CAS32503-27-8
IUPAC Nametetrabutylazanium hydrogen sulfate
Molecular FormulaC16H37NO4S
InChI KeySHFJWMWCIHQNCP-UHFFFAOYSA-M
SMILESOS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(10 % in water) Almost clear colorless
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point169°C to 173°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection