Thermo Scientific Chemicals

4-Hydroxymethylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 394340010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Hydroxymethylbenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 394340010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 394340010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3006-96-0
IUPAC Name
4-(hydroxymethyl)benzoic acid
Molecular Formula
C8H8O3
InChI Key
WWYFPDXEIFBNKE-UHFFFAOYSA-N
SMILES
OCC1=CC=C(C=C1)C(O)=O
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
181°C to 186°C
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text