2-Phenylthiazole, 98%, Thermo Scientific Chemicals
2-Phenylthiazole, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Phenylthiazole, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 394370050
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1826-11-5
IUPAC Name2-phenyl-1,3-thiazole
Molecular FormulaC9H7NS
InChI KeyWYKHSBAVLOPISI-UHFFFAOYSA-N
SMILESS1C=CN=C1C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Colorless to yellow
Appearance (Form)Liquid
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only