Thermo Scientific Chemicals

N,N-Diethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 395000500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

N,N-Diethyl-p-phenylenediamine sulfate, 99%, Thermo Scientific Chemicals

Catalog number: 395000500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 395000500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6283-63-2
IUPAC Name
N1,N1-diethylbenzene-1,4-diamine; sulfuric acid
Molecular Formula
C10H18N2O4S
InChI Key
AYLDJQABCMPYEN-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.CCN(CC)C1=CC=C(N)C=C1
Appearance (Color)
White to beige
Appearance (Form)
Crystalline powder or crystals and/or lumps
Infrared spectrum
Conforms
Melting point
184°C to 187°C
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text