Thermo Scientific Chemicals

(2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific Chemicals

Catalog number: 395020010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(2S,3S)-3-(N-BOC-amino)-1-oxirane-4-phenylbutane, 98%, Thermo Scientific Chemicals

Catalog number: 395020010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 395020010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
98737-29-2
IUPAC Name
tert-butyl N-[1-(oxiran-2-yl)-2-phenylethyl]carbamate
Molecular Formula
C15H21NO3
InChI Key
NVPOUMXZERMIJK-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C1CO1
Appearance (Color)
White to off-white
Appearance (Form)
Powder
Infrared spectrum
Conforms
HPLC
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text