Thermo Scientific Chemicals

Tetrabutylphosphonium hydroxide, 40 wt.% solution in water, Thermo Scientific Chemicals

Catalog number: 395031000
100 mL, Plastic Bottle, Each
Thermo Scientific Chemicals

Tetrabutylphosphonium hydroxide, 40 wt.% solution in water, Thermo Scientific Chemicals

Catalog number: 395031000
100 mL, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 395031000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14518-69-5, 7732-18-5
Appearance (Color)
Clear colorless to light yellow
Appearance (Form)
Solution
Titration with HCl
37.5 to 42.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text