Vinylboronic acid pinacol ester, 97+%, stabilized with 0.05% BHT, Thermo Scientific Chemicals
Vinylboronic acid pinacol ester, 97+%, stabilized with 0.05% BHT, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vinylboronic acid pinacol ester, 97+%, stabilized with 0.05% BHT, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 395530050
Price (USD)/ Each
176.65
Online exclusive
196.00 
Save 19.35 (10%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
176.65
Online exclusive
196.00 
Save 19.35 (10%)
Add to cart
Vinylboronic acid pinacol ester, 97+%, stabilized with 0.05% BHT, Thermo Scientific Chemicals
Catalog number395530050
Price (USD)/ Each
176.65
Online exclusive
196.00 
Save 19.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75927-49-0
IUPAC Name2-ethenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC8H15BO2
InChI KeyDPGSPRJLAZGUBQ-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to amber
GC>=97.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4240 to 1.4260 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only