Thermo Scientific Chemicals

Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid, Thermo Scientific Chemicals

Catalog number: 395620250
25 mL, Plastic bottle, Each
Thermo Scientific Chemicals

Titanium(III) chloride, 20% w/v solution in 2N Hydrochloric acid, Thermo Scientific Chemicals

Catalog number: 395620250
25 mL, Plastic bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 395620250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7705-07-9, 7647-01-0, 7732-18-5
Appearance (Color)
Dark purple
Appearance (Form)
Solution
Titration Complexometric
18 to 23 %
Titration with NaOH
1.5 to 2.5 N HCl

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text