2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals
2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Chloronicotinoyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 395790050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS49609-84-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Appearance (Form)Low melting solid
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Melting point39°C to 44°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only