Thermo Scientific Chemicals

Dichlorobis(tricyclohexylphosphine)palladium(II), 98%, Thermo Scientific Chemicals

Catalog number: 395890010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

Dichlorobis(tricyclohexylphosphine)palladium(II), 98%, Thermo Scientific Chemicals

Catalog number: 395890010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 395890010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
29934-17-6
IUPAC Name
dichloropalladium; bis(tricyclohexylphosphane)
Molecular Formula
C36H66Cl2P2Pd
InChI Key
VUYVXCJTTQJVKJ-UHFFFAOYSA-L
SMILES
Cl[Pd]Cl.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
Elemental Analysis
>=97.5 %
Appearance (Color)
Yellow
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text