1x1 image pixel for data collection
Potassium hydroxide, ca. 50% solution in water
Potassium hydroxide, ca. 50% solution in water
Potassium hydroxide, ca. 50% solution in water
Potassium hydroxide, ca. 50% solution in water
Thermo Scientific Chemicals

Potassium hydroxide, ca. 50% solution in water

CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3960600252.5 kgPlastic bottle
Catalog number 396060025
Price (USD)
102.00
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
102.00
Each
Add to cart
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Viscous solution
Titration with HCl49 to 51 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link