Thermo Scientific Chemicals

Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized, Thermo Scientific Chemicals

Catalog number: 396130010
1 L, Glass bottle, Each
Thermo Scientific Chemicals

Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized, Thermo Scientific Chemicals

Catalog number: 396130010
1 L, Glass bottle, Each
Quantity
Packaging
Catalog number: 396130010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5614-37-9
IUPAC Name
methoxycyclopentane
Molecular Formula
C6H12O
InChI Key
SKTCDJAMAYNROS-UHFFFAOYSA-N
SMILES
COC1CCCC1
Infrared spectrum
Conforms
Refractive index
1.4190 to 1.4220 (20°C, 589 nm)
Residue after evaporation
=<0.01 %
Appearance (Form)
Clear liquid
Peroxides
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text