N,N-Dimethylacetamide, 99.5%, Extra Dry, AcroSeal™
N,N-Dimethylacetamide, 99.5%, Extra Dry, AcroSeal™
N,N-Dimethylacetamide, 99.5%, Extra Dry, AcroSeal™
N,N-Dimethylacetamide, 99.5%, Extra Dry, AcroSeal™
N,N-Dimethylacetamide, 99.5%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

N,N-Dimethylacetamide, 99.5%, Extra Dry, AcroSeal™

CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 396351000
Price (USD)
76.90
Each
-
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
76.90
Each
Add to cart
Chemical Identifiers
CAS127-19-5
IUPAC NameN,N-dimethylacetamide
Molecular FormulaC4H9NO
InChI KeyFXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILESCN(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=99.4 %
Residue after evaporation=<0.005 %
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link