Thermo Scientific Chemicals

Lithium isopropoxide, 3M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 396501000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Lithium isopropoxide, 3M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 396501000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 396501000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2388-10-5
IUPAC Name
lithium(1+) propan-2-olate
Molecular Formula
C3H7LiO
InChI Key
HAUKUGBTJXWQMF-UHFFFAOYSA-N
SMILES
[Li+].CC(C)[O-]
Appearance (Color)
Clear yellow to red
Strength
2.9 to 3.3 M
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text